ContactUsLinkComponent
​Electric Von Frey Apparatus
​Electric Von Frey Apparatus
Catalog # 76670-398
Supplier:  MAZE ENGINEERS
​Electric Von Frey Apparatus
Catalog # 76670-398
Supplier:  MAZE ENGINEERS
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Electric Von Frey apparatus, mouse/rat
  • Cat. no.:
    76670-398
Specifications
About this item
The Von Frey monofilament was designed for the assessment of mechanical nociception.
  • Clear acrylic construction
  • Each chamber comes with two dividers
  • Chamber can hold 2 rats with one divider and 4 mice with 2 dividers
  • Chamber has a lid with air holes and the top is easy to open and close
The Von Frey monofilament was designed by Maximilian Von Frey as an esthesiometer in 1896. These filaments have been widely used in the assessment of mechanical nociception. The process of assessment of punctuating mechanical allodynia and hyperalgesia involves the application of Von Frey filaments of forces ranging anywhere from 0.08 mN to 2940 mN, and observing the withdrawal thresholds. The assay can be performed manually or by using an electric Von Frey apparatus. The electric device is used to overcome the disadvantages of the conventional assay. The former has many advantages over the manual assay, primarily the use of a single filament to apply varying levels of pressure. Further, unlike other tests such as the Randall-Selitto test, the Von Frey monofilament assay does not require restraining the subject. This approach minimizes handling and restraining anxiety which could otherwise potentially influence the performance of the subject.
Customers Who Bought This Also Bought