ContactUsLinkComponent
Tail Flick Test
Tail Flick Test
Catalog # 76670-288
Supplier:  MAZE ENGINEERS
Tail Flick Test
Catalog # 76670-288
Supplier:  MAZE ENGINEERS
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Tail flick test, mouse or rat
  • Cat. no.:
    76670-288
Specifications
About this item
The tail flick test was first described to assess analgesic effects of different substances on the nociceptive responses of rodents.
The Tail Flick test was first described by D’Amour and Smith in the year 1941 to assess analgesic effects of different substances on the nociceptive responses of rodents. The test is similar to the Hargreaves Plantar test, but instead of directing heat to the center of the rodent’s hind paw, heat stimulus is applied to the tail of the rodent.
Customers Who Bought This Also Bought