Spécifications
- Description:Static rods test; mouse/rat
- Material:Acrylic/metal
Spécifications
A propos de cet article
The static rods test is composed of dowels and clamps, and is used to assess motor coordination capabilities of the subject that can be affected by examples of pharmacological compounds, lesions, or diseases.
- Acrylic frame and metal rods
- Easy to use and setup
- Easy to clean assemble, and disassemble
Dowels of varying thicknesses are used to increase the difficulty of the task and refine the assessment further. One end of the dowel is attached to the edge of a fixture such that the rods horizontally protrude out in the space at a certain height from the ground.
Rod 1 is 35 mm dia., 60 cm length. Rod 2 is 28 mm dia., 60 cm length. Rod 3 is 22 mm dia., 60 cm length. Rod 4 is 15 mm dia., 60 cm length. Rod 5 is 9 mm dia., 60 cm length.