A propos de cet article
The vertical tank array is used for anxiety experiments for Zebrafish.
- Clear acrylic construction
- Each tank holds 1.5 L in a trapezoid pattern
- Three or six tank array formats
Vertical diving behavior is used to assess anxiety in zebrafish. Narrow tanks minimize horizontal movement to maximize the behavior assessment. Each tank holds 1.5 L in a trapezoid pattern. Sets of three or six arrays come together with a housing apparatus for easy video grading.