ContactUsLinkComponent
Circular Active Place Avoidance Arena
Circular Active Place Avoidance Arena
# de catalogue 76670-330
Fournisseur:  MAZE ENGINEERS
Circular Active Place Avoidance Arena
# de catalogue 76670-330
Fournisseur:  MAZE ENGINEERS
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Circular active place avoidance arena, mouse/rat
Spécifications
A propos de cet article
The APA is a circular metal arena shock grid underneath a rotating arena to study active and passive avoidance behaviors.
  • Includes grid floor, stainless steel base, rotating arena
  • Shock scrambler, shock cables, high precision serving motor also included
The subjects are allowed to explore the arena while avoiding a prohibited sector by using visual cues in and around the arena. Punishment in the form of shock is delivered to the subject if it enters the prohibited sector. Passive avoidance is observed when the arena is not rotating while active avoidance is observed when the arena is rotating. The arena makes the active place avoidance task suitable for evaluating spatial navigation performances of weanling rodents, making this an effective tool for investigating the effects of brain lesions, pharmacological manipulations, and diseases on spatial learning and memory.
Ordering information: Software package (see accessory option) is designed for use with the automated active place avoidance and video tracking software such as EthoVision or ANY-Maze.
Customers Who Bought This Also Bought