Body Turning Tests
Fournisseur: MAZE ENGINEERS
Certificats
A propos de cet article
The body turning test is a narrow alleyway that allows mice and rats to navigate turning or reversing.
- Acrylic construction
- Non reflective surfaces
- Removable walls for easy cleaning and storage
A narrow corridor open at one end and closed at the other (three walls), all resting on a removable base.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.