ContactUsLinkComponent
Digital Shaking Water Bath, 10 L
Digital Shaking Water Bath, 10 L
# de catalogue 470353-788
Fournisseur:  EDVOTEK
New Product
WATER BATH DIGITAL SHAKING 10L
Digital Shaking Water Bath, 10 L
# de catalogue 470353-788
Fournisseur:  EDVOTEK
New Product
Spécifications
  • Capacity:
    10 L
  • Description:
    Digital shaking water bath
  • Shaking speed:
    5 - 100 rpm
  • Cat. no.:
    470353-788
  • Interior Dimensions:
    30×22.5×15 cm
  • Electrical:
    110 V, 50 Hz, 300 W
  • Details:
    Shaking speed: 5 - 100 RPM
Spécifications
A propos de cet article
10 L Digital shaking water bath is designed for optimal sample incubation and samples are simultaneously shaken, heated and aerated.
  • Low water level automatic shut-off
  • Temperature range: Ambient to 99 °C (with cover)
  • Temperature accuracy: ±0.5 °C (with cover)
  • 2 year warranty
Edvotek®'s 10 L Digital Shaking Water Bath is designed for optimal sample incubation. Solutions are simultaneously shaken, heated and aerated to speed and improve reactions. Temperature control is from ambient to 75 or 99 °C with the optional cover.
Ordering information: Cover, available separately, helps to maintain temperature.
Customers Who Bought This Also Bought