Bumper Guards for Volumetric Flasks
Supplier: United Scientific Supplies
New Product
Certificates
About this item
Protect volumetric flasks from accidentally being tipped over.
- Wide design to cushion flasks if tipped over
- Made of high-quality silicone
Silicone bumpers slip easily onto any volumetric flask. In the event of a flask being accidentally tipped over, the wide bumpers cushion the neck from hitting the table surface. Color-coded and available in five different sizes.
Customers Who Bought This Also Bought