Magnetic Wands
# de catalogue 470353-638
Fournisseur: United Scientific Supplies
New Product
Spécifications
- Description:Magnetic wands
- Cat. no.:470353-638
Spécifications
A propos de cet article
Colorful wands to explore magnetism.
- Six wands in assorted colors
- Designed for simple magnetic activities
- Magnet encased in plastic
This easy-to-hold plastic wand contains a ceramic magnet. Use for various activities to learn what items are attracted to magnets. Each wand measures 8" long by 1" wide. Sold as a set of six magnets in assorted colors.
Customers Who Bought This Also Bought