Dissecting Trays with Wax
Fournisseur: United Scientific Supplies
New Product
Certificats
A propos de cet article
Dissection trays for all sizes of specimens.
- Aluminum construction
- Rounded corners include tie-down holes
- Wax holds dissection pins securely
Dissecting pans are made of durable aluminum and include a layer of wax. The rounded corners include holes that can be used as tie-down points to hold specimens securely during dissection.
Customers Who Bought This Also Bought