Graduated Cylinders, Double Scale, Borosilicate Glass, Class B
Fournisseur: United Scientific Supplies
New Product
Certificats
A propos de cet article
High-quality graduated cylinders with double scales.
- Made from heavy-duty borosilicate 3.3 glass
- Meets ASTM Class B tolerances for precision; individually calibrated to contain
- One-piece molded glass construction with hexagonal base and adjustable plastic bumper guard
- Low temperature gradient; can withstand high temperatures and thermal shocks
- Strain point: 515 °C
- Annealing point: 565 °C
- Softening point: 820 °C
Each cylinder features a double metric scale, calibrated to contain. Borosilicate glass cylinders with pour spouts feature heavy uniform wall tubing and attached glass hexagonal bases. All cylinders include bumper guards.
Certifications: Comply with ASTM E1272, Class B standards.
Customers Who Bought This Also Bought