Streak Plates, Glass
# de catalogue 470353-570
Fournisseur: United Scientific Supplies
New Product
Spécifications
- Description:Streak plates, glass
- L×W:2×1"
- Cat. no.:470353-570
Spécifications
A propos de cet article
Clear glass for mineral testing.
- ¹/₄" thick glass
- Overall dimensions: 2×1"
Designed for use in hardness testing of rocks and minerals.
Packaging: Plates are sold as packs of 10.
Customers Who Bought This Also Bought