A propos de cet article
Rabbit monoclonal [ARC1025] antibody to BMP6 for WB and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2,000, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: BMP6
Clonality: Monoclonal
Clone: ARC1025
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 77218-618
- Antigen name:
- BMP-6
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Clone:
- ARC1025
- Host:
- Rabbit
- Gene ID:
- UniprotID# P22004
- Isotype:
- IgG