Order Entry
ContactUsLinkComponent
Anti-Cyclin E1 Mouse Recombinant Antibody [clone: rCCNE1/4936]
Anti-Cyclin E1 Mouse Recombinant Antibody [clone: rCCNE1/4936]
# de catalogue 76952-250
Fournisseur:  ANTIBODIES.COM LLC
New Product
Anti-Cyclin E1 Mouse Recombinant Antibody [clone: rCCNE1/4936]
# de catalogue 76952-250
Fournisseur:  ANTIBODIES.COM LLC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Antibody type:
    Primary
  • Cat. no.:
    76952-250

Spécifications

A propos de cet article

Recombinant mouse monoclonal [rCCNE1/4936] antibody to Cyclin E1 for WB and IHC-P with samples derived from Human.

  • Validated Applications: WB, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cyclin E1
Clonality: Recombinant
Clone: rCCNE1/4936
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Customers Who Bought This Also Bought