To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Beta 4 tubulin
- Clonality:Monoclonal
- Clone:TUBB/4470
- Gene ID:UniprotID# P07437
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Antigen symbol:beta Tubulin
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:MGC117247|beta Tubulin|M40|beta Ib tubulin|Class I beta tubulin|Beta1 tubulin|MGC16435|Beta 5 tubulin|OK/SW cl.56
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:55 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant full-length human Tubulin beta 5 protein (TUBB5).
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Spécifications
A propos de cet article
Mouse monoclonal [TUBB/4470] antibody to beta Tubulin for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: beta Tubulin
Clonality: Monoclonal
Clone: TUBB/4470
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human