A propos de cet article
Rabbit polyclonal antibody to SAMM50 / SAM50 for WB, IHC and IP with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, IP
- Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200, IP: 1:500 to 1:1000
Type: Primary
Antigen: SAMM50/SAM50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 76866-560
- Antigen symbol:
- SAMM50/SAM50
- Antibody type:
- Primary
- Clonality:
- Polyclonal
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q9Y512
- Isotype:
- IgG