To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:BMP 15
- Clonality:Polyclonal
- Gene ID:UniprotID# O95972
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:BMP15
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:ODG2|BMP15_HUMAN|BMP-15|BMP15|GDF-9B|GDF 9B|Growth/differentiation factor 9B|GDF9B|Bone morphogenetic protein 15
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:45 kDa
- Storage temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from the Internal region of human BMP15 (amino acids 291 - 340)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Packaging:Plastic vial
Spécifications
A propos de cet article
Rabbit polyclonal antibody to BMP15 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: BMP15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human