Anti-CD307c Mouse Monoclonal Antibody [Clone: H5]
76965-186
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Host:
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Cat. no.:76965-186
Mouse monoclonal [H5] antibody to CD307c for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: CD307c
Clonality: Monoclonal
Clone: H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human