Order Entry
ContactUsLinkComponent
Anti-beta III Tubulin Rabbit Recombinant Antibody [Clone: TUBB3/7089R]
Anti-beta III Tubulin Rabbit Recombinant Antibody [Clone: TUBB3/7089R]
# de catalogue 76957-914
Fournisseur:  ANTIBODIES.COM LLC
New Product
ANTIBODY A277956-100 100UG
Anti-beta III Tubulin Rabbit Recombinant Antibody [Clone: TUBB3/7089R]
# de catalogue 76957-914
Fournisseur:  ANTIBODIES.COM LLC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Cat. no.:
    76957-914

Spécifications

A propos de cet article

Recombinant rabbit monoclonal [TUBB3/7089R] antibody to beta III Tubulin for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: beta III Tubulin
Clonality: Recombinant
Clone: TUBB3/7089R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought