Order Entry
ContactUsLinkComponent
Anti-HPV16 E1 + E4 Mouse Monoclonal Antibody [Clone: HPV16 E1/E4]
Anti-HPV16 E1 + E4 Mouse Monoclonal Antibody [Clone: HPV16 E1/E4]
# de catalogue 76966-374
Fournisseur:  ANTIBODIES.COM LLC
New Product
ANTIBODY A254182-100 100UG
Anti-HPV16 E1 + E4 Mouse Monoclonal Antibody [Clone: HPV16 E1/E4]
# de catalogue 76966-374
Fournisseur:  ANTIBODIES.COM LLC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Cat. no.:
    76966-374

Spécifications

A propos de cet article

Mouse monoclonal [HPV16 E1/E4] antibody to HPV16 E1 + E4 for IHC-P with samples derived from HPV-16.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: HPV16 E1 + E4
Clonality: Monoclonal
Clone: HPV16 E1/E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: HPV-16

Customers Who Bought This Also Bought