To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:BMP 15
- Clonality:Monoclonal
- Clone:BMP15/4263
- Gene ID:UniprotID# O95972
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:BMP15
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:ODG2|BMP15_HUMAN|BMP-15|BMP15|GDF-9B|GDF 9B|Growth/differentiation factor 9B|GDF9B|Bone morphogenetic protein 15
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:45 kDa
- Sequence:SAEVTASSSKHSGPENNQC
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:C terminal human GDF9B protein
- Purification:Protein A/G chromatography
- Packaging:Plastic vial
Spécifications
A propos de cet article
Mouse monoclonal [BMP15/4263] antibody to BMP15 for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, WB
- Recommended Dilutions: WB: 1 to 2 µg/ml
Type: Primary
Antigen: BMP15
Clonality: Monoclonal
Clone: BMP15/4263
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human