To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:alpha Tubulin
- Clonality:Monoclonal
- Clone:TUBA/3038
- Gene ID:UniprotID# P68363, P68366
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:alpha Tubulin
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:H2 ALPHA|Hum a tub2|ALS22|MGC171407|Hum a tub1|bA408E5.3|B ALPHA 1|Alpha-tubulin 1|LIS3
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:55 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant fragment, around amino acids 1-115, of human alpha Tubulin protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Packaging:Plastic vial
Spécifications
A propos de cet article
Mouse monoclonal [TUBA/3038] antibody to alpha Tubulin for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: alpha Tubulin
Clonality: Monoclonal
Clone: TUBA/3038
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human