Anti-CD27 Mouse Monoclonal Antibody [clone: LT27] (PerCP-Cyanine 5.5)
76949-882
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:CD 27
- Antigen symbol:CD27
- Clonality:Monoclonal
- Clone:LT27
- Conjugation:PerCP-Cyanine 5.5
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Size:100 tests
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P26842
- Antigen synonyms:CD27_HUMAN|S152|CD27 antigen|CD27 molecule|CD27L receptor|MGC20393|OTTHUMP00000238557|CD27|LPFS2
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Human peripheral blood lymphocytes
- Purification:This antibody is conjugated with tandem dye PerCP-Cyanine 5.5 under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. no.:76949-882
- Packaging:Glass bottle
Mouse monoclonal [LT27] antibody to CD27 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: LT27
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human