Anti-BMP-8 Rabbit Polyclonal Antibody
# de catalogue 77064-840
Fournisseur: ANTIBODIES.COM LLC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Clonality:Polyclonal
- Gene ID:UniprotID# P34820,Q7Z5Y6
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Form:Liquid
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:44 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide derived from human BMP-8 (amino acids 41-90)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Packaging:Plastic vial
Spécifications
A propos de cet article
Rabbit polyclonal antibody to BMP-8 for IHC and ELISA with samples derived from Human.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Customers Who Bought This Also Bought