To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:CCNE
- Clonality:Polyclonal
- Gene ID:UniprotID# P24864
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:Cyclin E1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:CyclinE|CCNE1_HUMAN|G1/S specific cyclin E|cyclin E variant ex7del|Ccne1|Cyclin Et|Cyclin E1|Cyclin Es|cyclin E variant ex5del
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:47 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Cyclin E1 (amino acids 43-92)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Packaging:Plastic vial
Spécifications
A propos de cet article
Rabbit polyclonal antibody to Cyclin E1 for IHC, IF and ELISA with samples derived from Human.
- Validated Applications: IHC, IF, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: Cyclin E1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human