The Flower of a Flowering Plant Microslides
Fournisseur: SUPERTEK SCIENTIFIC LLC
New Product
Certificats
A propos de cet article
Learn about flower anatomy and function.
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects It on both sides
- Set of 15 Includes detailed lesson pan on the topic under study
- Durable, inexpensive teaching resource
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery: Set Includes: 1. Complete flower (12×) 2. Stamens (18×) 3. Anther (18×) 4. Pistil (5×) 5. Ovary (15×) 6. Ovule (200×) 7. Pollen grain (480×) 8. Double fertilization - C.S. Stained (420×).
Customers Who Bought This Also Bought