The Cell Membrane Microslides
Fournisseur: SUPERTEK SCIENTIFIC LLC
New Product
Certificats
A propos de cet article
Explore the similarities and differences of various cell membranes.
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects It on both sides
- Set of 15 Includes detailed lesson pan on the topic under study
- Durable, inexpensive teaching resource
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery: Set Includes: 1. Epithelial cells (900×) 2. Cell membrane (3500×) 3. Paramecium-(dying) (200×) 4. Cilia -(Locomotion) (1500×) 5. Blood cells - (Structures) (26×) 6. Blood cells -(Functions) (600×) 7. White blood cell-(Recognition) (800×) 8. Onion skin cells (100×).
Customers Who Bought This Also Bought