The Kingdom Protista Microslides
Fournisseur: SUPERTEK SCIENTIFIC LLC
New Product
Certificats
A propos de cet article
Explore the Kingdom Protista.
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects It on both sides
- Set of 15 Includes detailed lesson pan on the topic under study
- Durable, inexpensive teaching resource
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery: Set Includes: 1. Amoeba/Actinophrys (200×) Phylum Sarcodina 2. Paramecium (100×) Phylum Ciliophora 3. Plasmodium Vivax (1000×) Phylum Apicomplexa 4. Trypanasoma Brucei Gambiense Smear - Stained (800×) Phylum Euglenozoa 5. Euglena (400×) Phylum Euglenapyta 6. Spirogyra Stained (175×) Phylum Charophyta 7. Noctiluca (63×) Phylum Dinoflagellata 8. Diatoms (50×) Phylum Heterokontophyta.
Customers Who Bought This Also Bought