Cytoplasm Microslides
Fournisseur: SUPERTEK SCIENTIFIC LLC
New Product
Certificats
A propos de cet article
Investigate the cytoplasm in different cells.
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
- Set of 15 includes detailed lesson pan on the topic under study
- Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery: Set includes 1. Cheek cells - stained (900×) 2. The modern cell -E.M. (37000×) 3. The rough endoplasmic reticulum - E.M. (18500×) 4. The smooth endoplasmic reticulum - E.M. (18500×) 5. The mitochondria - E.M. (105000×) 6. Golgi bodies - E.M. (9000×) 7. Vacuoles - E.M. (1800×) 8. Chloroplasts - E.M. (16000×).
Customers Who Bought This Also Bought