ContactUsLinkComponent
Luer Lock Syringes, 1 ml LDS, 2-Part, Non Sterile
Luer Lock Syringes, 1 ml LDS, 2-Part, Non Sterile
Catalog # 77232-932
Supplier:  Air-Tite
New Product
SYRINGE LUER LOCK 1ML 2PRT UNSTER BG100
Luer Lock Syringes, 1 ml LDS, 2-Part, Non Sterile
Catalog # 77232-932
Supplier:  Air-Tite
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    1 ml Luer lock, LDS, 2-part syringe
  • Cat. no.:
    77232-932
Specifications
About this item
An unsterile packaged 1 ml 2-part syringe with low dead space plunger for laboratory use.
  • 1 ml syrnge
  • 2-part laboratory syringe
  • LDS Low Dead Space
  • Plastic disposable syringe
  • Luer lock
  • All plastic
  • No rubber
  • No silicone oil
These 1 ml Luer lock syringes extend our popular 2-part syringe series. Part of the Air-Tite products premium laboratory brand, these syringes are made to our exacting specifications, comply with relevant hypodermic ISO requirements, and contain no rubber or silicone oil.
Packaging: Non sterile packaged in bulk convenience bags.
Customers Who Bought This Also Bought