A propos de cet article
Advanced, digital hotplate stirrers for all your laboratory heating and stirring needs.
- SafetyHeat™ early overtemperature detection system
- Rugged, spill-resistant metal housing
- Chemical-resistant ceramic top
- 2 years warranty
These hotplate stirrers are designed for economical heating and stirring in all laboratory settings. The bright LCD display includes easy-to-read temperature and speed settings, intuitive icons and a prominent hot top indicator that lights up when the heater is above 40 °C. Select from three plate sizes to suit your application. All models can be used with the optional temperature probe for precise temperature control of the sample.
Applications:
Boiling liquids, water/oil bath preparation, sample and solution preparation, dissolving buffers and reagents, preparing media, concentrating samples.
Display:
Backlit LCD Display for temperature and speed, green indicator lights when unit is heating and stirring, prominent hot top indicator light, probe in use icon, single point calibration icon.
Operation:
Stirring from 80 to 1600 rpm, ambient temperature range from +5 to 500 °C (ceramic top) or 380 °C (ceramic-coated steel top), power cord (included), stirring models include PTFE-coated stir bar.
Construction:
Durable metal housing, PET front panel, control knobs, on/off switch for heater disconnect, built-in support rod holder, easy-to-clean ceramic or ceramic-coated stainless steel tops.
Design Features:
Hot top indicator light to protect user; green indicator lights illuminate when unit is heating and stirring; temperature probe connector for use with optional temperature probe.
Spécifications
- Catalog No:
- 77270-244
- 77270-242
- 77270-240
- W×D×H:
- 264×163×110 mm
- 318×223×112 mm
- 264×163×101 mm