Scrubber for DK and DKL Series of Kjeldahl Digestion Units, KS 1000
Catalog # 77257-076
Supplier: VELP SCIENTIFIC INC.
New Product
Specifications
- Description:KS 1000 scrubber for DK and DKL series digestion units
- Cat. no.:77257-076
Specifications
About this item
Designed to neutralize toxic fumes produced during mineralization and safeguard laboratory operators without requiring any connection to tap water.
- Safeguard laboratory environment and operators
- Neutralizes corrosive toxic gases
- Quick and easy cleaning and maintenance
The KS 1000 Scrubber effectively neutralizes corrosive and toxic gases in two steps:
Dilution: In the right tank, reaction gases flow into a water solution, where they are effectively dissolved. This step allows even part of the sulphur dioxide produced during digestion to be trapped.
Neutralization: In the left glass tank, reaction gases are neutralised in a sodium hydroxide solution. Thanks to a wide contact surface between gas and liquid, the KS 1000 Scrubber achieves maximum neutralization efficiency.
Customers Who Bought This Also Bought