Eisco Alcohol Lamp
# de catalogue 470346-104
Fournisseur: Wards
Spécifications
- Description:Alcohol lamp
- Cat. no.:470346-104
Spécifications
A propos de cet article
Alcohol lamp with a polyhedral shape for positioning flame at different angles.
- Traditional alcohol lamp with 120 ml glass tank
- Features nickel-plated cap and fiber wick
- Polyhedral shape allows for positioning flame at multiple angles
- Use with 95% denatured alcohol as fuel
Glass alcohol lamp with metal cap and fiber wick has a 120 ml fuel capacity. Safer alternative to Bunsen and propane burners for basic laboratory applications. Sufficient for performing standard biology and sterilization procedures. Use with 95% denatured ethyl alcohol. Trim wick to no more than 2 cm.
Customers Who Bought This Also Bought