ContactUsLinkComponent
AccuTec Pro Hobby Knife and Blades
AccuTec Pro Hobby Knife and Blades
# de catalogue 77172-146
Fournisseur:  Accutec Blades
New Product
KNIFE HOBBY #1 W/5BLADES PK1
AccuTec Pro Hobby Knife and Blades
# de catalogue 77172-146
Fournisseur:  Accutec Blades
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    No.1 hobby knife and #11 hobby blades
Spécifications
A propos de cet article
AccuTec Pro No.1 hobby knife and #11 hobby blades are great for detailed cutting and trimming of lightweight materials.
  • Sure-grip barrel for comfort and control
  • Four jaw chuck
  • Heavy-duty knurled collar for durability
  • Plastic safety cap protects knife when not in use
This set includes five #11 blades in a storage tube. The hobby knife is compatible with #10, #11, #16, and #17 hobby blades.
Case dimensions: 10.44×7.88×9.75"
Packaging: Each card contains one hobby knife and five #11 hobby blades.
Caution: Blades are extremely sharp. Wear eye protection. Handle and use with care. Bending blades may break them. Keep out of children’s reach. Dispose of safely.
Customers Who Bought This Also Bought