SARS-CoV-2 (COVID-19) Membrane Peptide
# de catalogue 77120-676
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Gene ID:43740571
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:SARS-CoV-2
- Conjugation:Unconjugated
- Size:0.05 mg
- Storage conditions:Store SARS-CoV-2 (COVID-19) Membrane Peptide at −20 °C, stable for one year.
- Protein synonyms:SARS Matrix protein|SARS-CoV-2/SARS-CoV Matrix Antibody: SARS-CoV-2 Matrix protein|Membrane protein|M protein|E1 glycoprotein
- Protein/peptide name:M
- Concentration:200 µg/ml
- Formulation:Liquid. PBS pH 7.2 (10 mM NaH₂PO₄, 10 mM Na₂HPO₄, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide.
- Shipping temperature:Blue ice
- Tested applications:Blocking
Spécifications
A propos de cet article
Customers Who Bought This Also Bought