Hepatitis C Recombinant Envelope 1 Protein (from E. coli)
# de catalogue 77133-116
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Hepatitis C virus (HCV)
- Conjugation:Unconjugated
- Size:0.01 mg
- Storage conditions:Store in working aliquots at −20 °C. Avoid freeze/thaw cycles. When working with proteins care should be taken to keep recombinant protein at a cool and stable temperature.
- Protein/peptide name:E1
- Purity:~95% by SDS-PAGE
- Molecular weight:27 kDa
- Concentration:Batch dependent
- Formulation:Liquid. 25 mM Tris-HCl, 0.1% SDS, pH 8.3.
- Shipping temperature:Blue ice
- Tested applications:ELISA
Spécifications
A propos de cet article
Customers Who Bought This Also Bought