BMP5 Peptide
# de catalogue 77125-786
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Gene ID:653
- Packaging:Vial
- Protein/peptide type:Synthetic
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:Bone morphogenetic protein 5|BMP-5
- Protein/peptide name:BMP5
- Formulation:Lyophilized. BMP5 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
Spécifications
A propos de cet article
Customers Who Bought This Also Bought