BMP3 Peptide
# de catalogue 77121-342
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Gene ID:651
- Packaging:Vial
- Protein/peptide type:Synthetic
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:BMP3A|BMP-3A|Bone morphogenetic protein 3A|Bone morphogenetic protein 3|BMP-3
- Protein/peptide name:BMP3
- Formulation:Lyophilized. BMP3 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
Spécifications
A propos de cet article
Customers Who Bought This Also Bought