Anti-Membrane Protein Rabbit Polyclonal Antibody
# de catalogue 77125-436
Fournisseur: Prosci
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Membrane protein
- Clonality:Polyclonal
- Gene ID:43740571
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Antigen symbol:Membrane Protein
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.02 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:SARS Matrix protein|SARS-CoV-2/SARS-CoV Matrix Antibody: SARS-CoV-2 Matrix protein|Membrane protein|M protein|E1 glycoprotein
- Storage buffer:SARS-CoV-2 (COVID-19) Membrane Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:SARS-CoV-2 (COVID-19) Membrane antibody can be stored at 4...730 °C up to one year. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/ml
- Shipping temperature:Blue ice
- Immunogen:Anti-SARS-CoV-2 (COVID-19) Membrane antibody 9165 was raised against a peptide corresponding to 15 amino acids near the carboxyl terminus of SARS-CoV-2 (COVID-19) Membrane protein. The immunogen is located within the last 50 amino acids of SARS-CoV-2 (COVID-19) Membrane protein.
- Purification:SARS-CoV-2 (COVID-19) Membrane Antibody is affinity chromatography purified via peptide column.
- Packaging:Vial
Spécifications
A propos de cet article
Type: Primary
Antigen: Membrane Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Customers Who Bought This Also Bought