Anti-QSOX1 Rabbit Polyclonal Antibody
Catalogue # 77120-992
Fournisseur: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:quiescin Q6 sulfhydryl oxidase 1
- Antigen symbol:QSOX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:5768
- Antigen synonyms:Quiescin Q6 sulfhydryl oxidase 1|Sulfhydryl oxidase 1|QSCN6|Q6
- Modification:None
- Storage buffer:QSOX1 antibody is supplied in PBS containing 0.02% sodium azide.
- Storage temperature:QSOX1 antibody can be stored at 4...730 °C for three months and –20...+730 °C, stable for up to one year.
- Concentration:1 mg/ml
- Shipping temperature:20
- Immunogen:QSOX1 antibody was raised against a 17 amino acid peptide near the center of human QSOX1.The immunogen is located within amino acids 360-410 of QSOX1.
- Purification:QSOX1 antibody is affinity chromatography purified via peptide column.
- Cat. no.:77120-992
- Packaging:Vial
Spécifications
À propos de cet article
Type: Primary
Antigen: QSOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse