GATA1 Peptide
# de catalogue 77133-750
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Gene ID:2623
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:ERYF1|NFE1|GF-1|GF1|Erythroid transcription factor|Eryf1|XLANP|XLTT|NF-E1|XLTDA|GATA-1
- Protein/peptide name:GATA1
- Formulation:Lyophilized. GATA1 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
- Cat. no.:77133-750
Spécifications
A propos de cet article
Customers Who Bought This Also Bought