DUSP1 Peptide
# de catalogue 77131-498
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Gene ID:1843
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:HumanRatMouse
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:CL100|PTPN10|MAP kinase phosphatase 1|HVH1|MKP-1|Dual specificity protein phosphatase hVH1|VH1|Dual specificity protein phosphatase 1|MKP1
- Protein/peptide name:DUSP1
- Formulation:Lyophilized. DUSP1 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
- Cat. no.:77131-498
Spécifications
A propos de cet article
Customers Who Bought This Also Bought