BMPR1A Peptide
# de catalogue 77126-088
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Gene ID:657
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:Bone morphogenetic protein receptor type-1A|Activin receptor-like kinase 3|SKR5|BMP type-1A receptor|ALK3|10q23del|CD292|ACVRLK3
- Protein/peptide name:BMPR1A
- Formulation:Lyophilized. BMPR1A peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
Spécifications
A propos de cet article
Customers Who Bought This Also Bought