BAP Peptide
Catalogue # 77125-060
Fournisseur: Prosci
Numéro CAS:
Spécifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Synthetic
- Source:
- Species:Human
- Size:0.1 mg
- Environmentally Preferable:
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Gene ID:64374
- Protein synonyms:ULG5|MSS|BAP|Nucleotide exchange factor SIL1|BiP-associated protein
- Protein/peptide name:SIL1
- Formulation:Lyophilized. BAP peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:RT
- Tested applications:Blocking
- Cat. no.:77125-060