ALDH1A1 Peptide
# de catalogue 77132-786
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Gene ID:216
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:HumanRatMouse
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:RALDH 1|PUMB1|ALDC|RALDH1|HEL-S-53e|Retinal dehydrogenase 1|ALDH1|HEL-9|ALDH-E1|HEL12|ALDH11
- Protein/peptide name:ALDH1A1
- Formulation:Lyophilized. ALDH1A1 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
Spécifications
A propos de cet article
Customers Who Bought This Also Bought