PEX5 Peptide
# de catalogue 77132-364
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Gene ID:5830
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:peroxisomal biogenesis factor 5|peroxin-5|OTTHUMP00000238353|OTTHUMP00000238194|FLJ50721|PTS1-BP|OTTHUMP00000238195|OTTHUMP00000238354|OTTHUMP00000238192|PEX5|OTTHUMP00000238356|OTTHUMP00000238193|PTS1R|PTS1 receptor|FLJ50634|FLJ51948|PXR1
- Protein/peptide name:PEX5
- Formulation:Lyophilized. PEX5 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
- Cat. no.:77132-364
Spécifications
A propos de cet article
Customers Who Bought This Also Bought