Human Recombinant BMP-2 Protein (Animal Free) (from E. coli)
# de catalogue 77126-728
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Gene ID:650
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Conjugation:Unconjugated
- Size:2 µg
- Storage conditions:The recombinant protein is stable for at least 2 years from date of receipt at −20 °C. Reconstituted protein is stable for at least 3 months when stored in working aliquots with a carrier protein at −20 °C. As with any protein, exposing the recombinant protein to repeated freeze/thaw cycles is not recommended. When working with proteins care should be taken to keep recombinant protein at a cool and stable temperature.
- Biological activity:Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 0.5 - 1.0 µg/ml.
- Protein synonyms:Bone Morphogenetic Protein-2|BMP-2A
- Protein/peptide name:BMP2
- Purity:≥98% by SDS-PAGE gel and HPLC analyses
- Sequence:MQAKHKQRKR LKSSCKRHPL YVDFSDVGWN DWIVAPPGYH AFYCHGECPF PLADHLNSTN HAIVQTLVNS VNSKIPKACC VPTELSAISM LYLDENEKVV LKNYQDMVVE GCGCR
- Formulation:Lyophilized
- Shipping temperature:Room temperature
Spécifications
A propos de cet article
Customers Who Bought This Also Bought