Human, Mouse, Rat Recombinant BMP-7 Protein (from CHO cells)
# de catalogue 77123-978
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Gene ID:655
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:HumanRatMouse
- Conjugation:Unconjugated
- Size:2 µg
- Storage conditions:The recombinant protein is stable for at least 2 years from date of receipt at −20 °C. Reconstituted protein is stable for at least 3 months when stored in working aliquots with a carrier protein at −20 °C. As with any protein, exposing the recombinant protein to repeated freeze/thaw cycles is not recommended. When working with proteins care should be taken to keep recombinant protein at a cool and stable temperature.
- Biological activity:Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 0.02 - 0.04 µg/ml.
- Protein synonyms:Osteogenic Protein-1 (OP-1)|Bone Morphogenetic Protein-7
- Protein/peptide name:BMP7
- Purity:≥98% by SDS-PAGE gel and HPLC analyses
- Sequence:MANVAENSSS DQRQACKKHE LYVSFRDLGW QDWIIAPEGY AAYYCEGECA FPLNSYMNAT NHAIVQTLVH FINPETVPKP CCAPTQLNAI SVLYFDDSSN VILKKYRNMV VRACGCH
- Formulation:Lyophilized
- Shipping temperature:Room temperature
Spécifications
A propos de cet article
Customers Who Bought This Also Bought