Human, Mouse Recombinant GDF-5 Protein (from E. coli)
# de catalogue 77134-904
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Gene ID:8200
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:HumanMouse
- Conjugation:Unconjugated
- Size:10 µg
- Storage conditions:The recombinant protein is stable for at least 2 years from date of receipt at −20 °C. Reconstituted protein is stable for at least 3 months when stored in working aliquots with a carrier protein at −20 °C. As with any protein, exposing the recombinant protein to repeated freeze/thaw cycles is not recommended. When working with proteins care should be taken to keep recombinant protein at a cool and stable temperature.
- Biological activity:The ED50 was determined by its ability to induce alkaline phosphatase production by ATDC-5 chondrogenic cells is 1.0 - 2.0 µg/ml.
- Protein synonyms:BMP-14|Cartilage-Derived Morphogenetic Protein-1 (CDMP-1)|Growth/Differentiation Factor-5
- Protein/peptide name:GDF5
- Purity:≥98% by SDS-PAGE gel and HPLC analyses
- Sequence:APLATRQGKR PSKNLKARCS RKALHVNFKD MGWDDWIIAP LEYEAFHCEG LCEFPLRSHL EPTNHAVIQT LMNSMDPEST PPTCCVPTRL SPISILFIDS ANNVVYKQYE DMVVESCGCR
- Formulation:Lyophilized
- Shipping temperature:Room temperature
Spécifications
A propos de cet article
Customers Who Bought This Also Bought