Human Recombinant GDF-2 Protein (from HEK293 cells)
# de catalogue 77133-226
Fournisseur: Prosci
New Product
Numéro CAS:
Spécifications
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Conjugation:Unconjugated
- Size:0.05 mg
- Storage conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:BMP9|GDF-2|GDF2|BMP-9
- Protein/peptide name:GDF-2
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:12.1 kDa
- Sequence:Ser 320 - Arg 429
- Formulation:Lyophilized. ACN, TFA in Water
- Shipping temperature:Room temperature
- Tested applications:ELISA,WB
Spécifications
A propos de cet article
Customers Who Bought This Also Bought